Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300796.1 | NP_001287725.1 | 3 | 266 | ||
NM_001300797.1 | NP_001287726.1 | 3 | 274 | |||
NM_006325.4 | NP_006316.1 | 4 | 320 | |||
XM_017019772.1 | XP_016875261.1 | 4 | 435 | |||
XM_017019773.1 | XP_016875262.1 | 3 | 172 | |||
GenBank mRNA | AF052578.1 | - | 3 | 201 | ||
AF054183.1 | - | 4 | 291 | |||
AF501887.1 | - | 3 | 177 | |||
AK300934.1 | - | 3 | 195 | |||
AK311223.1 | - | 2 | 284 | |||
AK312466.1 | - | 4 | 243 | |||
AK316193.1 | - | 3 | 194 | |||
AL357616.1 | - | 4 | 230 | |||
BC000852.1 | - | 4 | 242 | |||
BC004272.1 | - | 4 | 205 | 3615048 | ||
BC014518.2 | - | 4 | 234 | 3830061 | ||
BC014901.2 | - | 4 | 220 | 3910005 | ||
BC016654.1 | - | 3 | 533 | 4342591 | ||
BC051908.2 | - | 4 | 245 | 5504629 | ||
BC072000.1 | - | 4 | 217 | 6644326 | ||
BT007271.1 | - | 3 | 177 | |||
CR450347.1 | - | 3 | 177 | |||
HY139187.1 | - | 4 | 316 | |||
M31469.1 | - | 3 | 177 |