Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030195.1 | - | 3 | 1190 | ||
AK014383.2 | - | 3 | 1066 | |||
AK045045.1 | - | 3 | 1200 | |||
BC017602.1 | - | 3 | 1142 | 4503004 | ||
BC089516.1 | - | 3 | 1352 | |||
BC103778.1 | - | 3 | 1134 | 6493935 |