Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023733.3 | NP_076222.1 | 15 | 1624 | ||
GenBank mRNA | AK004567.2 | - | 15 | 1625 | ||
BC006593.1 | - | 15 | 1612 | 3156501 | ||
BC012308.1 | - | 15 | 1639 | 3500167 |