Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205385.1 | NP_001192314.1 | 8 | 762 | ||
NM_001205386.1 | NP_001192315.1 | 8 | 759 | |||
NM_023735.2 | NP_076224.1 | 7 | 954 | |||
GenBank mRNA | AK004554.1 | - | 7 | 768 | ||
AK083343.1 | - | 7 | 772 | |||
AK145536.1 | - | 7 | 774 | |||
AK150487.1 | - | 7 | 656 | |||
AK150589.1 | - | 7 | 732 | |||
AK150991.1 | - | 7 | 712 | |||
AK152865.1 | - | 7 | 749 | |||
AK153079.1 | - | 7 | 775 | |||
AK153150.1 | - | 7 | 749 | |||
AK153204.1 | - | 7 | 655 | |||
AK159135.1 | - | 7 | 741 | |||
AK159668.1 | - | 7 | 666 | |||
AK159775.1 | - | 7 | 753 | |||
AK160047.1 | - | 7 | 614 | |||
AK166529.1 | - | 8 | 706 | |||
AK166586.1 | - | 8 | 706 | |||
AK167148.1 | - | 7 | 713 | |||
AK167891.1 | - | 7 | 711 | |||
AK168568.1 | - | 7 | 641 | |||
AK168771.1 | - | 7 | 727 | |||
AK187649.1 | - | 2 | 75 | |||
AK211935.1 | - | 3 | 192 | |||
BC005557.1 | - | 7 | 717 | 3156748 | ||
BC080806.1 | - | 8 | 720 | 5350752 | ||
BC108386.1 | - | 7 | 685 | 5051125 |