Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198836.1 | NP_001185765.1 | 1 | 368 | ||
NM_001198837.1 | NP_001185766.1 | 1 | 368 | |||
NM_006328.3 | NP_006319.1 | 1 | 368 | |||
GenBank mRNA | AF080561.1 | - | 1 | 309 | ||
AF315632.1 | - | 1 | 324 | |||
AF315633.1 | - | 1 | 324 | |||
AK021768.1 | - | 1 | 320 | |||
AK222830.1 | - | 1 | 321 | |||
BC000488.2 | - | 1 | 304 | 2819856 | ||
BC023619.2 | - | 1 | 304 | |||
DB044990.1 | - | 1 | 369 | |||
DQ294957.1 | - | 1 | 235 | |||
KU178520.1 | - | 1 | 229 | |||
KU178521.1 | - | 1 | 229 | |||
KU178522.1 | - | 1 | 229 |
Gene Symbol: RBM14-RBM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198845.1 | NP_001185774.1 | 1 | 368 | ||
NM_001198846.1 | NP_001185775.1 | 1 | 368 |