Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290448.1 | NP_001277377.1 | 8 | 1786 | ||
NM_001290449.1 | NP_001277378.1 | 8 | 1786 | |||
NM_023739.3 | NP_076228.2 | 8 | 1749 | |||
GenBank mRNA | AF223576.1 | - | 8 | 1713 | ||
AK005038.2 | - | 8 | 1721 | |||
AK033850.1 | - | 8 | 1680 | |||
AK050749.1 | - | 8 | 1723 | |||
AK144647.1 | - | 8 | 1735 | |||
AK146108.1 | - | 8 | 1703 | |||
AK155387.1 | - | 8 | 1701 | |||
AK160013.1 | - | 8 | 1719 | |||
BC053709.1 | - | 8 | 1684 | 30052746 |