Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 5 | 537 | ||
NM_001166548.1 | NP_001160020.1 | 5 | 541 | |||
NM_001166549.1 | NP_001160021.1 | 5 | 645 | |||
NM_023743.2 | NP_076232.2 | 5 | 552 | |||
GenBank mRNA | AB031388.1 | - | 5 | 373 | ||
AK041598.1 | - | 5 | 503 | |||
AK090363.1 | - | 5 | 443 | |||
AK128990.1 | - | 5 | 643 | |||
AK165678.1 | - | 5 | 540 | |||
BC033410.1 | - | 5 | 387 | 5359290 |