Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 19 | 3342 | ||
NM_001166548.1 | NP_001160020.1 | 18 | 3274 | |||
NM_001166549.1 | NP_001160021.1 | 18 | 3378 | |||
NM_023743.2 | NP_076232.2 | 19 | 3357 | |||
GenBank mRNA | AB031388.1 | - | 19 | 3178 | ||
AK012082.1 | - | 9 | 1761 | |||
AK041598.1 | - | 18 | 3236 | |||
AK088425.1 | - | 14 | 2444 | |||
AK128990.1 | - | 18 | 3374 | |||
AK165678.1 | - | 19 | 3344 | |||
BC033410.1 | - | 19 | 3192 | 5359290 |