Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023788.3 | NP_076277.1 | 1 | 785 | ||
GenBank mRNA | AF319981.1 | - | 1 | 560 | ||
AK008349.1 | - | 1 | 419 | |||
AK145866.1 | - | 1 | 747 | |||
BC051935.1 | - | 1 | 574 | 5714800 | ||
BC058380.1 | - | 1 | 564 | 6820349 | ||
BC060080.1 | - | 1 | 574 | 5714800 |