Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024239.2 | NP_077201.1 | 4 | 473 | ||
GenBank mRNA | AB010123.1 | - | 4 | 420 | ||
AK017600.1 | - | 4 | 428 | |||
AK019907.1 | - | 4 | 423 | |||
AK046995.1 | - | 4 | 425 | |||
AK136961.1 | - | 4 | 1024 | |||
AK139391.1 | - | 4 | 448 | |||
AK168810.1 | - | 4 | 446 | |||
AK220541.1 | - | 2 | 6704 | |||
BC003497.1 | - | 4 | 416 | 3153829 | ||
BC006939.1 | - | 4 | 471 | 2649718 | ||
BC025111.1 | - | 4 | 415 | 4194332 |