Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024243.4 | NP_077205.3 | 5 | 867 | ||
GenBank mRNA | AK002230.1 | - | 5 | 835 | ||
AK053323.1 | - | 5 | 812 | |||
AK089958.1 | - | 5 | 837 | |||
AK150526.1 | - | 5 | 802 | |||
AK150890.1 | - | 5 | 857 | |||
AK151258.1 | - | 5 | 835 | |||
AK151361.1 | - | 5 | 856 | |||
AK151879.1 | - | 5 | 856 | |||
AK151908.1 | - | 5 | 810 | |||
AK152070.1 | - | 5 | 810 | |||
AK165604.1 | - | 5 | 837 | |||
AK170503.1 | - | 5 | 847 | |||
BC003235.1 | - | 5 | 840 | 3497631 |