Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024245.4 | NP_077207.3 | 6 | 717 | ||
GenBank mRNA | AK013939.1 | - | 6 | 655 | ||
AK030771.1 | - | 6 | 694 | |||
AK076271.1 | - | 1 | 92 | |||
BC003363.1 | - | 6 | 690 | |||
BC047273.1 | - | 6 | 675 | 6440797 |