Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024254.3 | NP_077216.1 | 2, 3 | 271 | ||
GenBank mRNA | AK010654.1 | - | 2, 3 | 250 | ||
AK036861.1 | - | 2, 3 | 271 | |||
AK049599.1 | - | 2, 3 | 257 | |||
AK151464.1 | - | 2, 3 | 257 | |||
AK152570.1 | - | 2, 3 | 236 | |||
AK162183.1 | - | 2, 3 | 201 | |||
AK168639.1 | - | 2, 3 | 269 | |||
BC003216.1 | - | 2, 3 | 227 | 3157982 | ||
BC012402.1 | - | 2, 3 | 227 | 3157982 | ||
BC080661.1 | - | 2, 3 | 227 | 4460520 |
Gene Symbol: Slc12a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062099.1 | - | 4253 | 6844732 |