Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024254.3 | NP_077216.1 | 10 | 2600 | ||
GenBank mRNA | AK036861.1 | - | 10 | 2600 | ||
AK150905.1 | - | 3 | 1747 | |||
AK151464.1 | - | 10 | 2586 | |||
AK152570.1 | - | 10 | 2566 | |||
AK168639.1 | - | 10 | 2598 | |||
BC080661.1 | - | 10 | 2556 | 4460520 |