Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301367.1 | NP_001288296.1 | 3 | 375 | ||
NM_024255.3 | NP_077217.2 | 2 | 277 | |||
GenBank mRNA | AJ293845.1 | - | 3 | 298 | ||
AK011910.1 | - | 2 | 235 | |||
AK085899.1 | - | 2 | 246 | |||
AK136773.1 | - | 2 | 244 | |||
AK144948.1 | - | 2 | 193 | |||
AK145230.1 | - | 2 | 137 | |||
AY725196.1 | - | 2 | 134 | |||
BC002211.1 | - | 2 | 150 | 3490303 | ||
BY327952.1 | - | 2 | 278 |