Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301367.1 | NP_001288296.1 | 6 | 730 | ||
NM_024255.3 | NP_077217.2 | 5 | 632 | |||
GenBank mRNA | AK011910.1 | - | 5 | 590 | ||
AK085899.1 | - | 5 | 601 | |||
AK136773.1 | - | 5 | 599 | |||
AK144948.1 | - | 5 | 548 | |||
AK145230.1 | - | 5 | 492 | |||
AY725196.1 | - | 5 | 489 | |||
BC002211.1 | - | 5 | 505 | 3490303 |