Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024256.2 | NP_077218.1 | 2 | 326 | ||
GenBank mRNA | AB019523.1 | - | 2 | 205 | ||
AK048428.1 | - | 2 | 325 | |||
AK079723.1 | - | 1 | 625 | |||
AK140424.1 | - | 2 | 292 | |||
AK144402.1 | - | 2 | 345 | |||
AK146264.1 | - | 2 | 237 | |||
AK219513.1 | - | 2 | 99 | |||
BC002103.1 | - | 2 | 193 | 3485805 | ||
BC004038.1 | - | 2 | 326 | 3496170 | ||
BC012930.1 | - | 2 | 314 | 4500993 |