Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024256.2 | NP_077218.1 | 3 | 605 | ||
GenBank mRNA | AB019523.1 | - | 3 | 484 | ||
AK140424.1 | - | 3 | 571 | |||
AK144402.1 | - | 3 | 624 | |||
AK146264.1 | - | 3 | 516 | |||
BC002103.1 | - | 3 | 472 | 3485805 | ||
BC004038.1 | - | 3 | 605 | 3496170 | ||
BC012930.1 | - | 3 | 593 | 4500993 |