Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024258.2 | NP_077220.2 | 4 | 443 | ||
GenBank mRNA | AK004594.2 | - | 4 | 418 | ||
BC003278.1 | - | 4 | 372 | 3582180 |