Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024261.2 | NP_077223.2 | 3 | 408 | ||
GenBank mRNA | AK006776.1 | - | 3 | 418 | ||
AK134431.1 | - | 3 | 375 | |||
BC002139.1 | - | 3 | 393 | |||
BC099894.1 | - | 3 | 501 | |||
BC138908.1 | - | 3 | 368 | 8861930 | ||
BC145732.1 | - | 3 | 368 | 40131035 | ||
BU148062.1 | - | 3 | 393 |