Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024263.4 | NP_077225.4 | 8 | 1354 | ||
GenBank mRNA | AB040488.1 | - | 8 | 1274 | ||
AB052097.1 | - | 8 | 1232 | |||
AK004732.1 | - | 8 | 1237 | |||
BC002140.1 | - | 3 | 351 | |||
BC026438.1 | - | 8 | 1276 | 4239626 | ||
EU849596.1 | - | 8 | 1192 | |||
EU849597.1 | - | 8 | 1183 |