Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026865.2 | NP_081141.1 | 6 | 1273 | ||
GenBank mRNA | AK007198.1 | - | 6 | 1272 | ||
BC057970.1 | - | 7 | 633 |