Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289535.1 | NP_001276464.1 | 6 | 626 | ||
NM_001289536.1 | NP_001276465.1 | 5 | 548 | |||
NM_026959.3 | NP_081235.2 | 6 | 626 | |||
GenBank mRNA | AK007700.1 | - | 6 | 624 | ||
AK016910.1 | - | 6 | 568 | |||
AK169273.1 | - | 6 | 570 | |||
AK170165.1 | - | 6 | 622 | |||
BC021362.1 | - | 5 | 538 | 5035350 |