Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026998.3 | NP_081274.2 | 4, 5 | 408 | ||
GenBank mRNA | AK011640.1 | - | 5, 6 | 410 | ||
AK013158.1 | - | 4, 5 | 409 | |||
AK182704.1 | - | 3, 4 | 347 | |||
BC025911.1 | - | 4, 5 | 395 | |||
BC061028.1 | - | 5, 6 | 449 | 6594585 |