Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027022.4 | NP_081298.1 | 1 | 240 | ||
GenBank mRNA | AK005603.1 | - | 1 | 193 | ||
AK006874.1 | - | 1 | 241 | |||
AK161303.1 | - | 1 | 234 | |||
AY162281.1 | - | 1 | 121 | |||
AY172740.1 | - | 1 | 121 | |||
AY241871.1 | - | 1 | 240 | |||
BC049747.1 | - | 1 | 233 | 6772661 |