Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027436.3 | NP_081712.2 | 4 | 634 | ||
GenBank mRNA | AK017491.1 | - | 4 | 610 | ||
AK038479.1 | - | 4 | 628 | |||
AK044460.1 | - | 4 | 619 | |||
AK049000.1 | - | 4 | 575 | |||
AK076613.1 | - | 4 | 562 | |||
AK146428.1 | - | 5 | 625 | |||
BC110358.1 | - | 4 | 612 | 5322591 | ||
BC144798.1 | - | 4 | 615 | |||
BC145711.1 | - | 4 | 615 | 40131014 | ||
CA460475.1 | - | 4 | 671 |