Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038479.1 | - | 19 | 2959 | ||
AK044460.1 | - | 15 | 2559 | |||
BC059092.1 | - | 1 | 263 | |||
BC144798.1 | - | 19 | 2946 | |||
BC145711.1 | - | 19 | 2946 | 40131014 |