Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027444.3 | NP_081720.2 | 5 | 617 | ||
GenBank mRNA | AF276950.1 | - | 5 | 502 | ||
AF276951.1 | - | 5 | 589 | |||
AF276952.1 | - | 2 | 345 | |||
AF454943.1 | - | 5 | 610 | |||
AF454944.1 | - | 3 | 482 | |||
AK017487.1 | - | 5 | 610 | |||
AK029532.1 | - | 3 | 484 | |||
AK029747.1 | - | 6 | 731 | |||
AK049516.1 | - | 5 | 603 | |||
AK050488.1 | - | 5 | 556 | |||
AK157813.1 | - | 5 | 596 | |||
AK167724.1 | - | 5 | 748 | |||
BC019456.1 | - | 5 | 692 | |||
BC024449.1 | - | 3 | 465 | |||
BC038874.1 | - | 1 | 156 | |||
BC057869.1 | - | 5 | 585 |