Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027444.3 | NP_081720.2 | 11 | 1364 | ||
GenBank mRNA | AF276950.1 | - | 11 | 1249 | ||
AF276951.1 | - | 11 | 1336 | |||
AF276952.1 | - | 8 | 1092 | |||
AF454943.1 | - | 11 | 1357 | |||
AF454944.1 | - | 9 | 1229 | |||
AK029532.1 | - | 9 | 1231 | |||
AK029747.1 | - | 12 | 1478 | |||
AK049516.1 | - | 11 | 1350 | |||
AK050488.1 | - | 11 | 1303 | |||
AK157813.1 | - | 11 | 1343 | |||
AK167724.1 | - | 11 | 1495 | |||
BC019456.1 | - | 11 | 1439 | |||
BC024449.1 | - | 9 | 1212 | |||
BC038874.1 | - | 7 | 903 | |||
BC057869.1 | - | 11 | 1332 | |||
BC066821.1 | - | 9 | 991 | 30061364 |