Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 9 | 1472 | ||
GenBank mRNA | AK017523.1 | - | 10 | 1908 | ||
AK045166.1 | - | 1 | 1294 | |||
AK075579.1 | - | 7 | 1772 | |||
AK154071.1 | - | 9 | 1460 | |||
BC010777.1 | - | 9 | 1472 | 4241357 |