Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027457.4 | NP_081733.3 | 3, 4 | 374 | ||
GenBank mRNA | AK009851.1 | - | 3, 4 | 357 | ||
AK043276.1 | - | 3, 4 | 342 | |||
AK158107.1 | - | 3, 4 | 353 | |||
BC019420.1 | - | 3, 4 | 338 | 5053733 |