Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027462.4 | NP_081738.2 | 2 | 161 | ||
GenBank mRNA | AK017602.1 | - | 2 | 172 | ||
AK033774.1 | - | 159 | ||||
AK036902.1 | - | 2 | 181 | |||
AK042715.1 | - | 2 | 162 | |||
AK042964.1 | - | 2 | 162 | |||
BC089583.1 | - | 2 | 183 | 6762967 |