Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5730508B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027482.3 | NP_081758.1 | 2 | 319 | ||
GenBank mRNA | AK012470.1 | - | 2 | 290 | ||
AK017758.1 | - | 2 | 282 | |||
AK162420.1 | - | 2 | 296 | |||
AK163003.1 | - | 2 | 286 | |||
BC039568.1 | - | 2 | 46 | |||
BC116791.1 | - | 2 | 266 | 40126110 | ||
BC116793.1 | - | 2 | 266 | 40126112 | ||
BC144728.1 | - | 3 | 297 |