Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 4 | 913 | ||
GenBank mRNA | AB055406.1 | - | 4 | 945 | ||
AK018128.1 | - | 4 | 729 | |||
AK036605.1 | - | 4 | 914 | |||
AK042557.1 | - | 4 | 626 | |||
AK047657.1 | - | 4 | 970 | |||
AK086092.1 | - | 4 | 922 | |||
AK138268.1 | - | 4 | 612 | |||
AK160904.1 | - | 4 | 920 | |||
AK161526.1 | - | 4 | 922 | |||
AK162725.1 | - | 4 | 914 | |||
AK163617.1 | - | 4 | 612 | |||
AK165529.1 | - | 4 | 710 | |||
BC026658.1 | - | 4 | 676 | |||
BC116246.1 | - | 4 | 835 | 40093530 | ||
BC116247.1 | - | 4 | 833 | 40093536 |