Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099735.1 | NP_001093205.1 | 7 | 915 | ||
NM_001099736.1 | NP_001093206.1 | 7 | 919 | |||
NM_001825.2 | NP_001816.2 | 8 | 1079 | |||
GenBank mRNA | AK122769.1 | - | 7 | 879 | ||
AK313293.1 | - | 7 | 906 | |||
BC029140.1 | - | 7 | 896 | 5195322 | ||
CR450315.1 | - | 6 | 837 | |||
J05401.1 | - | 7 | 1035 |