Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293791.1 | NP_001280720.1 | 3 | 350 | ||
NM_001293792.1 | NP_001280721.1 | 3 | 350 | |||
NM_027590.4 | NP_081866.1 | 3 | 350 | |||
GenBank mRNA | AK014939.1 | - | 3 | 326 | ||
AK030974.1 | - | 3 | 313 | |||
AK034498.1 | - | 3 | 266 | |||
AK040574.1 | - | 3 | 351 | |||
AK047093.1 | - | 3 | 345 | |||
AK083331.1 | - | 3 | 305 | |||
AK147743.1 | - | 3 | 324 |