Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027591.3 | NP_081867.1 | 7 | 651 | ||
NM_029378.2 | NP_083654.1 | 5 | 465 | |||
GenBank mRNA | AK014934.1 | - | 7 | 652 | ||
BC147175.1 | - | 7 | 623 | 9056223 | ||
BC147176.1 | - | 7 | 623 | 9056216 | ||
DQ316772.1 | - | 7 | 607 | |||
DQ316773.1 | - | 5 | 465 |