Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027597.4 | NP_081873.2 | 4 | 611 | ||
NM_029069.1 | NP_083345.1 | 4 | 616 | |||
GenBank mRNA | AB473726.1 | - | 2 | 291 | ||
AK014851.1 | - | 4 | 617 | |||
AK015802.1 | - | 4 | 463 | |||
BC051149.1 | - | 4 | 471 | 6707726 |