Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027597.4 | NP_081873.2 | 5, 6 | 1264 | ||
NM_029069.1 | NP_083345.1 | 5, 6 | 1269 | |||
GenBank mRNA | AB473726.1 | - | 3, 4 | 944 | ||
AK014851.1 | - | 5, 6 | 1270 | |||
AK015802.1 | - | 5, 6 | 1116 | |||
BC051149.1 | - | 5, 6 | 1124 | 6707726 |