Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr20rt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027614.1 | NP_081890.1 | 1 | 796 | ||
GenBank mRNA | AK015014.1 | - | 1 | 796 | ||
AK015210.1 | - | 246 | ||||
BC050778.1 | - | 1 | 583 | 6743775 |