Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105530.1 | NP_001099000.1 | 3 | 354 | ||
NM_006367.3 | NP_006358.1 | 3 | 372 | |||
XM_005270368.1 | XP_005270425.1 | 3 | 212 | |||
XM_011540509.1 | XP_011538811.1 | 4 | 765 | |||
XM_011540510.1 | XP_011538812.1 | 4 | 762 | |||
XM_011540511.1 | XP_011538813.1 | 3 | 196 | |||
XM_011540512.1 | XP_011538814.1 | 3 | 315 | |||
XM_011540513.2 | XP_011538815.1 | 3 | 207 | |||
XM_011540514.1 | XP_011538816.1 | 4 | 322 | |||
XM_011540515.1 | XP_011538817.1 | 4 | 762 | |||
XM_017000067.1 | XP_016855556.1 | 3 | 280 | |||
XM_017000068.1 | XP_016855557.1 | 4 | 324 | |||
XM_017000069.1 | XP_016855558.1 | 4 | 535 | |||
XM_017000070.1 | XP_016855559.1 | 3 | 554 | |||
XM_017000071.1 | XP_016855560.1 | 3 | 512 | |||
XM_017000072.1 | XP_016855561.1 | 3 | 448 | |||
XM_017000073.1 | XP_016855562.1 | 3 | 269 | |||
GenBank mRNA | AK096193.1 | - | 3 | 206 | ||
AK222513.1 | - | 3 | 213 | |||
AK295396.1 | - | 4 | 134 | |||
AK297829.1 | - | 3 | 204 | |||
AK298110.1 | - | 3 | 156 | |||
AK300861.1 | - | 3 | 210 | |||
AK315731.1 | - | 3 | 259 | |||
BC013963.2 | - | 3 | 189 | 2963922 | ||
BC095440.1 | - | 3 | 194 | 30370256 | ||
BT007152.1 | - | 2 | 143 | |||
CR457409.1 | - | 2 | 143 | |||
L12168.1 | - | 2 | 182 | |||
M98474.1 | - | 3 | 213 |