Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027654.2 | NP_081930.1 | 3 | 583 | ||
GenBank mRNA | AB047007.1 | - | 3 | 633 | ||
AK016709.1 | - | 3 | 605 | |||
AK132448.1 | - | 3 | 629 | |||
AK155067.1 | - | 3 | 644 | |||
BC016195.1 | - | 3 | 583 | 4488614 | ||
BC089460.1 | - | 3 | 622 | 30632537 |