Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177881.1 | NP_001171352.1 | 3 | 1684 | ||
NM_001177882.1 | NP_001171353.1 | 3 | 1452 | |||
NM_001293769.1 | NP_001280698.1 | 2 | 1146 | |||
NM_027756.5 | NP_082032.1 | 3 | 1576 | |||
GenBank mRNA | AK016959.1 | - | 3 | 1453 | ||
AK089654.1 | - | 2 | 1147 | |||
AK122332.1 | - | 2 | 1033 | |||
AK133407.1 | - | 3 | 1685 | |||
BC095950.1 | - | 3 | 1522 | 5025281 |