Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNCRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159674.1 | NP_001153146.1 | 2, 3 | 262 | ||
NM_001159675.1 | NP_001153147.1 | 1, 2 | 168 | |||
NM_001159676.1 | NP_001153148.1 | 1, 2 | 168 | |||
NM_001159677.1 | NP_001153149.1 | 2, 3 | 697 | |||
NM_006372.4 | NP_006363.4 | 2, 3 | 697 | |||
XM_005248635.4 | XP_005248692.1 | 2, 3 | 318 | |||
XM_005248637.2 | XP_005248694.1 | 2, 3 | 281 | |||
XM_017010176.1 | XP_016865665.1 | 2, 3 | 318 | |||
XM_017010177.1 | XP_016865666.1 | 2, 3 | 318 | |||
XM_017010178.1 | XP_016865667.1 | 2, 3 | 278 | |||
XM_017010179.1 | XP_016865668.1 | 2, 3 | 253 | |||
GenBank mRNA | AF037448.1 | - | 2, 3 | 681 | ||
AF155568.1 | - | 2, 3 | 360 | |||
AK222776.1 | - | 2, 3 | 306 | |||
AK313572.1 | - | 2, 3 | 317 | |||
AY034481.1 | - | 1, 2 | 156 | |||
AY034482.1 | - | 1, 2 | 156 | |||
AY034483.1 | - | 1, 2 | 156 | |||
BC015575.2 | - | 2, 3 | 238 | |||
BC024283.1 | - | 2, 3 | 265 | |||
BC032643.1 | - | 2, 3 | 253 | 5495201 |