Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027769.2 | NP_082045.1 | 7, 8 | 746 | ||
GenBank mRNA | AK017651.1 | - | 7, 8 | 751 | ||
AK036664.1 | - | 7, 8 | 725 | |||
AK146324.1 | - | 7, 8 | 723 | |||
AK220348.1 | - | 7, 8 | 717 | |||
BC036983.1 | - | 7, 8 | 673 | |||
BC049226.1 | - | 5, 6 | 507 | |||
BC090632.1 | - | 6, 7 | 647 | 30620886 | ||
BY717432.1 | - | 7, 8 | 746 |