Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082924.1 | - | 3 | 846 | ||
AK159067.1 | - | 3 | 2259 | |||
BC096370.1 | - | 3 | 686 | 3597940 | ||
BC131971.1 | - | 2 | 656 | 40130248 | ||
BC131975.1 | - | 2 | 656 | 40130252 |