Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141950.1 | NP_001135422.1 | 6 | 1160 | ||
NM_027797.3 | NP_082073.1 | 6 | 1229 | |||
GenBank mRNA | AK017460.1 | - | 6 | 1229 | ||
AK049405.1 | - | 2 | 1090 | |||
AK134821.1 | - | 6 | 1161 | |||
AK169083.1 | - | 6 | 1119 | |||
BC022756.1 | - | 6 | 1104 | 4922223 |