Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289686.1 | NP_001276615.1 | 5 | 374 | ||
NM_001289687.1 | NP_001276616.1 | 2 | 245 | |||
NM_001289688.1 | NP_001276617.1 | 2 | 419 | |||
NM_027871.2 | NP_082147.1 | 3 | 219 | |||
GenBank mRNA | AK004600.2 | - | 3 | 190 | ||
AK029872.1 | - | 6 | 404 | |||
BC005517.1 | - | 2 | 216 | 3494309 | ||
BC012262.1 | - | 2 | 197 | 4192378 | ||
BM963858.1 | - | 2 | 419 | |||
BY224523.1 | - | 3 | 219 | |||
CD539300.1 | - | 2 | 235 |