Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163645.1 | NP_001157117.1 | 5 | 820 | ||
NM_027881.3 | NP_082157.2 | 4 | 430 | |||
GenBank mRNA | AK004768.1 | - | 4 | 432 | ||
AK037114.1 | - | 4 | 430 | |||
AK160310.1 | - | 5 | 821 | |||
AK173006.1 | - | 5 | 629 | |||
BC044866.1 | - | 4 | 465 | 5003631 | ||
BC086485.1 | - | 5 | 732 | 5344992 |