Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYOU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130991.2 | NP_001124463.1 | 3 | 247 | ||
NM_006389.4 | NP_006380.1 | 3 | 380 | |||
XM_005271392.3 | XP_005271449.1 | 3 | 241 | |||
XM_005271393.3 | XP_005271450.1 | 3 | 221 | |||
XM_005271394.3 | XP_005271451.1 | 3 | 288 | |||
XM_017017095.1 | XP_016872584.1 | 3 | 276 | |||
XM_017017096.1 | XP_016872585.1 | 3 | 297 | |||
XM_017017097.1 | XP_016872586.1 | 3 | 220 | |||
GenBank mRNA | AK222756.1 | - | 3 | 273 | ||
AK294905.1 | - | 4 | 408 | |||
AK297260.1 | - | 3 | 206 | |||
AK299648.1 | - | 3 | 203 | |||
AK314178.1 | - | 3 | 271 | |||
BC004560.2 | - | 3 | 195 | |||
BC017726.1 | - | 3 | 198 | |||
BC072436.1 | - | 3 | 246 | 5756154 | ||
BP236985.1 | - | 3 | 215 | |||
BP358970.1 | - | 3 | 339 | |||
DC364982.1 | - | 3 | 339 | |||
DQ350134.1 | - | 3 | 268 | |||
DQ372932.1 | - | 3 | 268 | |||
HY089857.1 | - | 3 | 248 | |||
U65785.1 | - | 3 | 248 |